missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-Mercaptohexyl Acetate, TCI America™

$61.11 - $207.78

Chemical Identifiers

CAS 136954-20-6
Molecular Formula C8H16O2S
Molecular Weight (g/mol) 176.274
MDL Number MFCD00792516
InChI Key JUCARGIKESIVLB-UHFFFAOYSA-N
Synonym Acetic Acid 3-Mercaptohexyl Ester
PubChem CID 518810
ChEBI CHEBI:77818
IUPAC Name 3-sulfanylhexyl acetate
SMILES CCCC(CCOC(=O)C)S
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
M24975G View Documents TCI America
M24975G
5g Each for $61.11
 
M249725G View Documents TCI America
M249725G
25g Each for $207.78
 
Specifications

Chemical Identifiers

136954-20-6
176.274
JUCARGIKESIVLB-UHFFFAOYSA-N
518810
3-sulfanylhexyl acetate
C8H16O2S
MFCD00792516
Acetic Acid 3-Mercaptohexyl Ester
CHEBI:77818
CCCC(CCOC(=O)C)S

Specifications

Colorless
136954-20-6
MFCD00792516
JUCARGIKESIVLB-UHFFFAOYSA-N
3-sulfanylhexyl acetate
518810
176.27
≥97.0% (GC)
5g
C8H16O2S
Acetic Acid 3-Mercaptohexyl Ester
CCCC(CCOC(=O)C)S
176.274
CHEBI:77818
Clear Liquid at 20°C
3-Mercaptohexyl Acetate
Safety and Handling

Safety and Handling

TSCA : Yes