missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-Mercapto-4-methyl-4H-1,2,4-triazole, TCI America™

$116.11 - $724.19

Chemical Identifiers

CAS 24854-43-1
Molecular Formula C3H5N3S
Molecular Weight (g/mol) 115.154
MDL Number MFCD00010069
InChI Key AGWWTUWTOBEQFE-UHFFFAOYSA-N
Synonym 3-mercapto-4-methyl-4h-1,2,4-triazole, 4-methyl-4h-1,2,4-triazole-3-thiol, 3-mercapto-4-methyl-1,2,4-triazole, 4-methyl-1,2,4-triazole-3-thiol, 3h-1,2,4-triazole-3-thione, 2,4-dihydro-4-methyl, 1,4-dihydro-4-methyl-3h-1,2,4-triazole-3-thione, 4-methyl-1,2,4-triazole-3-thione, 2,4-dimethyl-4-methyl-3h-1,2,4-triazole-3-thione, 4-methyl-4h-1,2,4 triazole-3-thiol, 3h-1,2,4-triazole-3-thione, 1,4-dihydro-4-methyl
PubChem CID 1256162
IUPAC Name 4-methyl-1H-1,2,4-triazole-5-thione
SMILES CN1C=NNC1=S
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
M122425G View Documents TCI America
M122425G
25g Each for $116.11
 
M1224250G View Documents TCI America
M1224250G
250g Each for $724.19
 
Specifications

Chemical Identifiers

24854-43-1
115.154
AGWWTUWTOBEQFE-UHFFFAOYSA-N
1256162
CN1C=NNC1=S
C3H5N3S
MFCD00010069
3-mercapto-4-methyl-4h-1,2,4-triazole, 4-methyl-4h-1,2,4-triazole-3-thiol, 3-mercapto-4-methyl-1,2,4-triazole, 4-methyl-1,2,4-triazole-3-thiol, 3h-1,2,4-triazole-3-thione, 2,4-dihydro-4-methyl, 1,4-dihydro-4-methyl-3h-1,2,4-triazole-3-thione, 4-methyl-1,2,4-triazole-3-thione, 2,4-dimethyl-4-methyl-3h-1,2,4-triazole-3-thione, 4-methyl-4h-1,2,4 triazole-3-thiol, 3h-1,2,4-triazole-3-thione, 1,4-dihydro-4-methyl
4-methyl-1H-1,2,4-triazole-5-thione

Specifications

White-Yellow
25g
C3H5N3S
3-mercapto-4-methyl-4h-1,2,4-triazole, 4-methyl-4h-1,2,4-triazole-3-thiol, 3-mercapto-4-methyl-1,2,4-triazole, 4-methyl-1,2,4-triazole-3-thiol, 3h-1,2,4-triazole-3-thione, 2,4-dihydro-4-methyl, 1,4-dihydro-4-methyl-3h-1,2,4-triazole-3-thione, 4-methyl-1,2,4-triazole-3-thione, 2,4-dimethyl-4-methyl-3h-1,2,4-triazole-3-thione, 4-methyl-4h-1,2,4 triazole-3-thiol, 3h-1,2,4-triazole-3-thione, 1,4-dihydro-4-methyl
CN1C=NNC1=S
115.154
115.15
≥98.0% (HPLC,T)
167°C
24854-43-1
MFCD00010069
AGWWTUWTOBEQFE-UHFFFAOYSA-N
4-methyl-1H-1,2,4-triazole-5-thione
1256162
Crystal-Powder at 20°C
3-Mercapto-4-methyl-4H-1,2,4-triazole
Safety and Handling

Safety and Handling

RTECSNumber : XZ5359126

TSCA : Yes