Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Mercapto-4-methyl-1,2,4-triazole, 97%

$111.35 - $352.58

Chemical Identifiers

Chemical Name or Material 3-Mercapto-4-methyl-1,2,4-triazole
CAS 24854-43-1
Molecular Formula C3H5N3S
Formula Weight 115.16
MDL Number MFCD00010069
Beilstein 110580
Synonym 4-Methyl-3-mercapto-1,2,4-triazole; 4-Methyl-1,2,4-triazole-3-thiol
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0800718

  SDS
alfa aesar™
L0800718
50g Each for $111.35

AAL0800730

  SDS
alfa aesar™
L0800730
250g Each for $352.58
Description & Specifications

Specifications

Formula Weight 115.16
Chemical Name or Material 3-Mercapto-4-methyl-1,2,4-triazole
TSCA TSCA
MDL Number MFCD00010069
CAS 24854-43-1
Beilstein 110580
Melting Point 166° to 169°C
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Assay 97%
Molecular Formula C3H5N3S
Synonym 4-Methyl-3-mercapto-1,2,4-triazole; 4-Methyl-1,2,4-triazole-3-thiol
RTECS Number XZ5359126
Health Hazard 1 Warning
Quantity 50g
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.