missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
3-Mercapto-1,2-propanediol, 90 wt% aqueous solution, ACROS Organics™
Manufacturer: Thermo Fisher Scientific AC125525000
Specifications
3-Mercapto-1, 2-propanediol | |
>112°C | |
Glass bottle | |
1.23 | |
Colorless to Yellow | |
96-27-5,7732-18-5 | |
92.0,12.0 | |
HSCH2CH(OH)CH2OH | |
01,519 | |
1-thioglycerol, 3-mercapto-1,2-propanediol, 3-mercaptopropane-1,2-diol, alpha-thioglycerol, glycerol-1-thiol, monothioglycerol, thioglycerin, thioglycerine, thioglycerol, thiovanol | |
PJUIMOJAAPLTRJ-UHFFFAOYSA-N | |
3-sulfanylpropane-1,2-diol | |
7291 | |
108.16 | |
≥88% |
1.2300g/mL | |
Authentic | |
1.5073 | |
118.0°C (5.0 mmHg) | |
500mL | |
88.0,8.0 | |
C3H8O2S | |
MFCD00004879 | |
15,9488 | |
Solubility in water: slightly soluble. Other solubilities: miscible with alcohol,insoluble in ether | |
C(C(CS)O)O | |
108.16 | |
CHEBI:74537 | |
Viscous Solution |
Chemical Identifiers
96-27-5 | |
108.16 | |
PJUIMOJAAPLTRJ-UHFFFAOYSA-N | |
7291 | |
3-sulfanylpropane-1,2-diol |
C3H8O2S | |
MFCD00004879 | |
1-thioglycerol, 3-mercapto-1,2-propanediol, 3-mercaptopropane-1,2-diol, alpha-thioglycerol, glycerol-1-thiol, monothioglycerol, thioglycerin, thioglycerine, thioglycerol, thiovanol | |
CHEBI:74537 | |
C(C(CS)O)O |
Safety and Handling
GHS H Statement
Harmful if swallowed.
Toxic in contact with skin.
Causes skin irritation.
May cause an allergic skin reaction.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF ON SKIN: Wash with plenty of soap and water.
If skin irritation or rash occurs: Get
GHS Signal Word: Danger
EINECSNumber : 202-495-0
RTECSNumber : TY8140000
TSCA : TSCA