Learn More
3-Mercapto-1,2-propanediol, 90%
CAS: 96-27-5 | C3H8O2S | 108.16 g/mol
Supplier: Thermo Scientific Chemicals A1894818
Description
It generally used as a size-regulating capping agent for nanocrystals. 1-Thioglycerol is also used in bacteriology experiments to induce the synthesis of porphyrin compounds in aerobically growing Escherichia coli. Also used to study the pH-sensitive photoluminescence of aqueous thiol-capped CdTe nanocrystals. 1-Thioglycerol is used to develop and test thiol functionalized copolymers. 1-Thioglycerol is used as a post-modification agent in the generation of non-standard peptide foldamers.Potential substitute for 2-mercaptoethanol, probe for the study of lymphocyte activation.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsIt generally used as a size-regulating capping agent for nanocrystals. 1-Thioglycerol is also used in bacteriology experiments to induce the synthesis of porphyrin compounds in aerobically growing Escherichia coli. Also used to study the pH-sensitive photoluminescence of aqueous thiol-capped CdTe nanocrystals. 1-Thioglycerol is used to develop and test thiol functionalized copolymers. 1-Thioglycerol is used as a post-modification agent in the generation of non-standard peptide foldamers.Potential substitute for 2-mercaptoethanol, probe for the study of lymphocyte activation.
Solubility
Slightly soluble in water and benzene. Soluble in acetone. Insoluble in ether.
Notes
Air Sensitive and Hygroscopic, store away from water/moisture and air. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store under dry inert gas.
Specifications
| 3-Mercapto-1,2-propanediol | |
| 1.247 | |
| 149°C (300°F) | |
| 1.511 | |
| MFCD00004879 | |
| 1732046 | |
| 14,9335 | |
| Slightly soluble in water and benzene. Soluble in acetone. Insoluble in ether. | |
| OCC(O)CS | |
| 108.16 | |
| CHEBI:74537 | |
| ≥90% |
| 96-27-5 | |
| 117°C to 119°C (5 mmHg) | |
| C3H8O2S | |
| HOCH2CH(OH)CH2SH | |
| 50 g | |
| Air sensitive; Hygroscopic | |
| 3-mercapto-1,2-propanediol, monothioglycerol, 1-thioglycerol, thioglycerol, thioglycerine, thioglycerin, thiovanol, 3-mercaptopropane-1,2-diol, alpha-thioglycerol, glycerol-1-thiol | |
| PJUIMOJAAPLTRJ-UHFFFAOYNA-N | |
| 3-sulfanylpropane-1,2-diol | |
| 7291 | |
| 108.16 |
Chemical Identifiers
| 96-27-5 | |
| 108.16 | |
| PJUIMOJAAPLTRJ-UHFFFAOYNA-N | |
| 7291 | |
| 3-sulfanylpropane-1,2-diol |
| C3H8O2S | |
| MFCD00004879 | |
| 3-mercapto-1,2-propanediol, monothioglycerol, 1-thioglycerol, thioglycerol, thioglycerine, thioglycerin, thiovanol, 3-mercaptopropane-1,2-diol, alpha-thioglycerol, glycerol-1-thiol | |
| CHEBI:74537 | |
| OCC(O)CS |
Safety and Handling
GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P272-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P333+P313-P361-P363-P501c
H302-H311-H315-H317-H319-H335
DOTInformation : Hazard Class: 6.1; Packaging Group: III
EINECSNumber : 202-495-0
RTECSNumber : TY8140000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only