Learn More
3-Maleimidopropionic acid N-hydroxysuccinimide ester, 99%, Thermo Scientific Chemicals
$310.25 - $951.03
Chemical Identifiers
CAS | 55750-62-4 |
---|---|
Molecular Formula | C11H10N2O6 |
Molecular Weight (g/mol) | 266.209 |
MDL Number | MFCD00043141 |
InChI Key | JKHVDAUOODACDU-UHFFFAOYSA-N |
Synonym | n-succinimidyl 3-maleimidopropionate, 2,5-dioxopyrrolidin-1-yl 3-2,5-dioxo-2,5-dihydro-1h-pyrrol-1-yl propanoate, 3-maleimido propionic acid n-hydroxysuccinimide ester, bmps, 3-maleimidopropionic acid n-succinimidyl ester, 3-maleimidopropionic acid nhs, succinimido 3-maleimidopropanoate, 2,5-dioxopyrrolidin-1-yl 3-2,5-dioxopyrrol-1-yl propanoate, 3-maleimidopropionic acid n-hydroxysuccinimide ester |
PubChem CID | 4620597 |
IUPAC Name | (2,5-dioxopyrrolidin-1-yl) 3-(2,5-dioxopyrrol-1-yl)propanoate |
SMILES | C1CC(=O)N(C1=O)OC(=O)CCN2C(=O)C=CC2=O |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAH27329MD
|
Thermo Scientific Chemicals
H27329MD |
250 mg |
Each for $310.25
|
|
|||||
AAH2732903
|
Thermo Scientific Chemicals
H2732903 |
1 g |
Each for $951.03
|
|
|||||
Description
3-Maleimidopropionic acid N-hydroxysuccinimide ester is a reagent that is widely used for generating stable maleimide-activated carrier proteins that can spontaneously react with sulfhydryls. Alternatively these relatively stable maleimide-activated intermediates may be lyophilized and stored for later conjugation to a hapten.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications3-Maleimidopropionic acid N-hydroxysuccinimide ester is a reagent that is widely used for generating stable maleimide-activated carrier proteins that can spontaneously react with sulfhydryls. Alternatively these relatively stable maleimide-activated intermediates may be lyophilized and stored for later conjugation to a hapten.
Solubility
Insoluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, heat, water, moisture.
Chemical Identifiers
55750-62-4 | |
266.209 | |
JKHVDAUOODACDU-UHFFFAOYSA-N | |
4620597 | |
C1CC(=O)N(C1=O)OC(=O)CCN2C(=O)C=CC2=O |
C11H10N2O6 | |
MFCD00043141 | |
n-succinimidyl 3-maleimidopropionate, 2,5-dioxopyrrolidin-1-yl 3-2,5-dioxo-2,5-dihydro-1h-pyrrol-1-yl propanoate, 3-maleimido propionic acid n-hydroxysuccinimide ester, bmps, 3-maleimidopropionic acid n-succinimidyl ester, 3-maleimidopropionic acid nhs, succinimido 3-maleimidopropanoate, 2,5-dioxopyrrolidin-1-yl 3-2,5-dioxopyrrol-1-yl propanoate, 3-maleimidopropionic acid n-hydroxysuccinimide ester | |
(2,5-dioxopyrrolidin-1-yl) 3-(2,5-dioxopyrrol-1-yl)propanoate |
Specifications
55750-62-4 | |
C11H10N2O6 | |
250 mg | |
n-succinimidyl 3-maleimidopropionate, 2,5-dioxopyrrolidin-1-yl 3-2,5-dioxo-2,5-dihydro-1h-pyrrol-1-yl propanoate, 3-maleimido propionic acid n-hydroxysuccinimide ester, bmps, 3-maleimidopropionic acid n-succinimidyl ester, 3-maleimidopropionic acid nhs, succinimido 3-maleimidopropanoate, 2,5-dioxopyrrolidin-1-yl 3-2,5-dioxopyrrol-1-yl propanoate, 3-maleimidopropionic acid n-hydroxysuccinimide ester | |
JKHVDAUOODACDU-UHFFFAOYSA-N | |
(2,5-dioxopyrrolidin-1-yl) 3-(2,5-dioxopyrrol-1-yl)propanoate | |
4620597 | |
99% |
168°C to 170°C | |
MFCD00043141 | |
Moisture sensitive | |
Insoluble in water. | |
C1CC(=O)N(C1=O)OC(=O)CCN2C(=O)C=CC2=O | |
266.209 | |
266.21 | |
3-Maleimidopropionic acid N-hydroxysuccinimide ester |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P271-P304+P340-P312-P501c
H335
TSCA : No
Recommended Storage : Keep cold
RUO – Research Use Only