missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-Maleimidopropiohydrazide Trifluoroacetate, TCI America™

$406.39 - $1,217.66

Chemical Identifiers

CAS 359436-61-6
Molecular Formula C9H10F3N3O5
Molecular Weight (g/mol) 297.19
MDL Number MFCD01863410
InChI Key BADCXPKRBUEEMA-UHFFFAOYSA-N
Synonym 3-Maleimidopropionic Hydrazide Trifluoroacetate
PubChem CID 71607324
IUPAC Name 3-(2,5-dioxopyrrol-1-yl)propanehydrazide;2,2,2-trifluoroacetic acid
SMILES C1=CC(=O)N(C1=O)CCC(=O)NN.C(=O)(C(F)(F)F)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
M27031G View Documents TCI America
M27031G
1g Each for $1,217.66
 
M2703250MG View Documents TCI America
M2703250MG
250mg Each for $406.39
 
Specifications

Chemical Identifiers

359436-61-6
297.19
BADCXPKRBUEEMA-UHFFFAOYSA-N
71607324
C1=CC(=O)N(C1=O)CCC(=O)NN.C(=O)(C(F)(F)F)O
C9H10F3N3O5
MFCD01863410
3-Maleimidopropionic Hydrazide Trifluoroacetate
3-(2,5-dioxopyrrol-1-yl)propanehydrazide;2,2,2-trifluoroacetic acid

Specifications

White
1g
C9H10F3N3O5
3-Maleimidopropionic Hydrazide Trifluoroacetate
C1=CC(=O)N(C1=O)CCC(=O)NN.C(=O)(C(F)(F)F)O
297.19
297.19
≥98.0% (HPLC)
127°C
359436-61-6
MFCD01863410
BADCXPKRBUEEMA-UHFFFAOYSA-N
3-(2,5-dioxopyrrol-1-yl)propanehydrazide;2,2,2-trifluoroacetic acid
71607324
Crystal-Powder at 20°C
3-Maleimidopropiohydrazide Trifluoroacetate
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Freezer (-20 °C)