missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-(Isopropylamino)propanol, TCI America™

$132.92 - $397.23

Chemical Identifiers

CAS 33918-15-9
Molecular Formula C6H15NO
Molecular Weight (g/mol) 117.192
MDL Number MFCD00483086
InChI Key GZCPWFOPXIDRDP-UHFFFAOYSA-N
PubChem CID 2063457
IUPAC Name 3-(propan-2-ylamino)propan-1-ol
SMILES CC(C)NCCCO
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
I075525G View Documents TCI America
I075525G
25g Each for $397.23
 
I07555G View Documents TCI America
I07555G
5g Each for $132.92
 
Specifications

Chemical Identifiers

33918-15-9
117.192
GZCPWFOPXIDRDP-UHFFFAOYSA-N
3-(propan-2-ylamino)propan-1-ol
C6H15NO
MFCD00483086
2063457
CC(C)NCCCO

Specifications

94°C
25g
C6H15NO
GZCPWFOPXIDRDP-UHFFFAOYSA-N
3-(propan-2-ylamino)propan-1-ol
2063457
Crystal-Lump at 20°C
3-(Isopropylamino)propanol
White
33918-15-9
MFCD00483086
CC(C)NCCCO
117.192
117.19
≥98.0% (GC,T)
Safety and Handling

Safety and Handling

TSCA : No