Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Isopropoxypropylamine, 98+%

Chemical Identifiers

Chemical Name or Material 3-Isopropoxypropylamine
CAS 2906-12-9
Molecular Formula (CH3)2CHO(CH2)3NH2
Formula Weight 117.19
MDL Number MFCD00008220
UN Number UN2733
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB23748AD

 
alfa aesar™
B23748AD
50mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAB23748AK

 
alfa aesar™
B23748AK
250mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 117.19
Chemical Name or Material 3-Isopropoxypropylamine
TSCA TSCA
Assay ≥98%
Molecular Formula (CH3)2CHO(CH2)3NH2
MDL Number MFCD00008220
CAS 2906-12-9
UN Number UN2733
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H314-H318-H226
Causes severe skin burns and eye damage.
Causes serious eye damage.
Flammable liquid and vapour.
Health Hazard 3 GHS P Statement
P210-P260-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: AMINES, FLAMMABLE, CORROSIVE, N.O.S.
Flash Point 42°C (108°F)
Boiling Point 149° to 151°C
Quantity 50mL
Safety and Handling

Danger

GHS P Statement
P210-P260-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H314-H318-H226
Causes severe skin burns and eye damage.
Causes serious eye damage.
Flammable liquid and vapour.