Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Isocyanatobenzoyl chloride, 97%

Chemical Identifiers

Chemical Name or Material 3-Isocyanatobenzoyl chloride
CAS 5180-79-0
Molecular Formula C8H4ClNO2
Formula Weight 181.58
MDL Number MFCD00013864
UN Number UN2922
Beilstein 2090273
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1600203

 
alfa aesar™
L1600203
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAL1600206

 
alfa aesar™
L1600206
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 181.58
Chemical Name or Material 3-Isocyanatobenzoyl chloride
MDL Number MFCD00013864
Health Hazard 1 Danger
Molecular Formula C8H4ClNO2
CAS 5180-79-0
UN Number UN2922
Beilstein 2090273
Odor Pungent
Health Hazard 2 GHS H Statement
H301-H331-H334-H314-H318
Toxic if swallowed.
Toxic if inhaled.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Health Hazard 3 GHS P Statement
P260-P301+P310-P303+P361+P353-P305+P351+P338-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Boiling Point 91°C (3mmHg)
Flash Point >110°C (230°F)
Recommended Storage 2° to 8°C
DOT Information Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUIDS, TOXIC, N.O.S.
Quantity 1g
Assay 97%
Safety and Handling

Danger

GHS P Statement
P260-P301+P310-P303+P361+P353-P305+P351+P338-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301-H331-H334-H314-H318
Toxic if swallowed.
Toxic if inhaled.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.

2° to 8°C