Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Isobutylpyridine, 99%

$86.66 - $86.66

Chemical Identifiers

Chemical Name or Material 3-Isobutylpyridine
CAS 14159-61-6
Molecular Formula C9H13N
Formula Weight 135.21
MDL Number MFCD02683082
Synonym 3-(2-Methylpropyl)pyridine
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2208088

  SDS
alfa aesar™
B2208088
2.5g Each for $86.66
Description & Specifications

Specifications

Formula Weight 135.21
Chemical Name or Material 3-Isobutylpyridine
TSCA TSCA
Assay 99%
Molecular Formula C9H13N
MDL Number MFCD02683082
CAS 14159-61-6
Synonym 3-(2-Methylpropyl)pyridine
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Quantity 2.5g
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.