Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Iodothiophene, 96%, stab. with copper

$87.43 - $295.54

Chemical Identifiers

Chemical Name or Material 3-Iodothiophene
CAS 10486-61-0
Name Note stabilized with copper
Molecular Formula C4H3IS
Formula Weight 210.04
MDL Number MFCD00037901
Beilstein 105451
Synonym 3-Thienyl iodide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1758103

  SDS
alfa aesar™
L1758103
1g Each for $87.43

AAL1758106

  SDS
alfa aesar™
L1758106
5g Each for $295.54
Description & Specifications

Specifications

Formula Weight 210.04
Chemical Name or Material 3-Iodothiophene
MDL Number MFCD00037901
CAS 10486-61-0
Beilstein 105451
Health Hazard 2 GHS H Statement
H315-H319-H227
Causes skin irritation.
Causes serious eye irritation.
Combustible liquid.
Health Hazard 3 GHS P Statement
P210-P280-P305+P351+P338-P362-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Take off contaminated clothing and wash before reuse.
Store in a well-ventilated place. Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Name Note stabilized with copper
Assay 96%
Molecular Formula C4H3IS
Synonym 3-Thienyl iodide
Quantity 1g
Flash Point 78°C (172°F)
Health Hazard 1 Warning
Boiling Point 77°C (11mmHg)
Safety and Handling

Warning

GHS P Statement
P210-P280-P305+P351+P338-P362-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Take off contaminated clothing and wash before reuse.
Store in a well-ventilated place. Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H227
Causes skin irritation.
Causes serious eye irritation.
Combustible liquid.