Please login to your online account to display your discounted pricing

3-Iodophenylboronic acid, 97%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 3-Iodophenylboronic acid
CAS 221037-98-5
CAS Min % 96.0
CAS Max % 100.0
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C6H6BIO2
Formula Weight 247.82
MDL Number MFCD01319015
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC35947EA

 
fisher scientific company
AC35947EA
Amber glass bottle 10g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥96.0%
Physical Form Powder
Quantity 10g
Color Beige
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Chemical Name or Material 3-Iodophenylboronic acid
MDL Number MFCD01319015
Health Hazard 2 GHS H Statement:
Causes serious eye irritation.
Harmful if swallowed.
Causes skin irritation.
May cause respiratory irritation.
Assay Percent Range 97%
Molecular Formula C6H6BIO2
Formula Weight 247.82
CAS Min % 96.0
CAS 221037-98-5
CAS Max % 100.0
Packaging Amber glass bottle
Infrared Spectrum Authentic