missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-Iodophenyl Isothiocyanate 98.0+%, TCI America™

$88.88 - $261.69

Chemical Identifiers

CAS 3125-73-3
Molecular Formula C7H4INS
Molecular Weight (g/mol) 261.08
MDL Number MFCD00041074
InChI Key LKCSXVAACHUTGC-UHFFFAOYSA-N
Synonym 1-iodanyl-3-isothiocyanato-benzene, 3-iodophenyl isothiocyanate, 3-iodophenylisothiocyanate, 3-ipi, acmc-1cmge, benzene, 1-iodo-3-isothiocyanato, benzene,1-iodo-3-isothiocyanato, isothiocyanic acid, m-iodophenyl ester, m-iodophenylisothiocyanate
PubChem CID 151083
IUPAC Name 1-iodo-3-isothiocyanatobenzene
SMILES C1=CC(=CC(=C1)I)N=C=S
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
I09851G View Documents Tci America
I09851G
1g Each for $88.88
 
I09855G View Documents Tci America
I09855G
5g Each for $261.69
 
Specifications

Chemical Identifiers

3125-73-3
261.08
LKCSXVAACHUTGC-UHFFFAOYSA-N
151083
C1=CC(=CC(=C1)I)N=C=S
C7H4INS
MFCD00041074
1-iodanyl-3-isothiocyanato-benzene, 3-iodophenyl isothiocyanate, 3-iodophenylisothiocyanate, 3-ipi, acmc-1cmge, benzene, 1-iodo-3-isothiocyanato, benzene,1-iodo-3-isothiocyanato, isothiocyanic acid, m-iodophenyl ester, m-iodophenylisothiocyanate
1-iodo-3-isothiocyanatobenzene

Specifications

White-Yellow
1g
C7H4INS
2923
LKCSXVAACHUTGC-UHFFFAOYSA-N
1-iodo-3-isothiocyanatobenzene
151083
Crystal-Lump at 20°C
3-Iodophenyl Isothiocyanate
47°C
3125-73-3
MFCD00041074
1-iodanyl-3-isothiocyanato-benzene, 3-iodophenyl isothiocyanate, 3-iodophenylisothiocyanate, 3-ipi, acmc-1cmge, benzene, 1-iodo-3-isothiocyanato, benzene,1-iodo-3-isothiocyanato, isothiocyanic acid, m-iodophenyl ester, m-iodophenylisothiocyanate
C1=CC(=CC(=C1)I)N=C=S
261.08
261.08
≥98.0% (GC)
Safety and Handling

Safety and Handling

TSCA : No