missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Iodophenyl isothiocyanate, 98%

Manufacturer:  Alfa Aesar™ L1023506

 View more versions of this product

Catalog No. AAL1023506



Specifications

Specifications

3-Iodophenyl isothiocyanate
44°C to 47°C
3125-73-3
MFCD00041074
2802538
LKCSXVAACHUTGC-UHFFFAOYSA-N
1-iodo-3-isothiocyanatobenzene
151083
98%
Pungent
5g
C7H4INS
UN2923
3-iodophenyl isothiocyanate, 3-iodophenylisothiocyanate, benzene, 1-iodo-3-isothiocyanato, m-iodophenylisothiocyanate, 3-ipi, isothiocyanic acid, m-iodophenyl ester, acmc-1cmge, benzene,1-iodo-3-isothiocyanato, 3-iodophenyl isothiocyanate, 1-iodanyl-3-isothiocyanato-benzene
C1=CC(=CC(=C1)I)N=C=S
261.08
261.09

Chemical Identifiers

3125-73-3
261.08
LKCSXVAACHUTGC-UHFFFAOYSA-N
151083
C1=CC(=CC(=C1)I)N=C=S
C7H4INS
MFCD00041074
3-iodophenyl isothiocyanate, 3-iodophenylisothiocyanate, benzene, 1-iodo-3-isothiocyanato, m-iodophenylisothiocyanate, 3-ipi, isothiocyanic acid, m-iodophenyl ester, acmc-1cmge, benzene,1-iodo-3-isothiocyanato, 3-iodophenyl isothiocyanate, 1-iodanyl-3-isothiocyanato-benzene
1-iodo-3-isothiocyanatobenzene
Safety and Handling

Safety and Handling

GHS H Statement
H331-H314-H318-H302-H312
Toxic if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Harmful in contact with skin.

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLIDS, TOXIC, N.O.S.

Documents