Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Iodophenyl isocyanate, 97%

Chemical Identifiers

Chemical Name or Material 3-Iodophenyl isocyanate
CAS 23138-56-9
Molecular Formula C7H4INO
Formula Weight 245.02
MDL Number MFCD00037088
UN Number UN2206
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH3080303

  SDS
alfa aesar™
H3080303
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH3080306

  SDS
alfa aesar™
H3080306
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 245.02
CAS 23138-56-9
UN Number UN2206
Health Hazard 2 GHS H Statement
H331-H334-H302-H315-H319-H335
Toxic if inhaled.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Recommended Storage 2° to 8°C
Odor Pungent
Boiling Point 55°C (0.2mmHg)
Quantity 1g
Molecular Formula C7H4INO
DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: ISOCYANATES, TOXIC, N.O.S.
Health Hazard 1 Danger
Flash Point >110°C (230°F)
MDL Number MFCD00037088
Chemical Name or Material 3-Iodophenyl isocyanate
Assay 97%
Health Hazard 3 GHS P Statement
P260-P280h-P305+P351+P338-P309-P310
Do not breathe dust/fume/gas/mist/vapors/spray.
Wear protective gloves/clothing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
Safety and Handling

Danger

GHS P Statement
P260-P280h-P305+P351+P338-P309-P310
Do not breathe dust/fume/gas/mist/vapors/spray.
Wear protective gloves/clothing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.

GHS H Statement
H331-H334-H302-H315-H319-H335
Toxic if inhaled.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

2° to 8°C