missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-Iodophenol, 99%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC249700050

 View more versions of this product

Catalog No. AC249700050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Specifications

Specifications

3-Iodophenol
Authentic
Beige-Yellow to Gray
5g
626-02-8
100.0
C6H5IO
MFCD00002261
Solubility in water: slightly soluble. Other solubilities: soluble in aqueous alkali,soluble in most common organic solvents
C1=CC(=CC(=C1)I)O
220
CHEBI:33439
Crystalline Powder, Crystals and/or Chunks
113°C
Glass bottle
38.0°C to 43.0°C
99%
98.5
98.5% min. (GC)
IC6H4OH
3-iodo-phenol, 3-jodphenol, 3-jodphenol german, 5-iodophenol, m-hydroxyiodobenzene, m-iodophenol, m-jodphenol, phenol, 3-iodo, phenol, m-iodo, pubchem3968
FXTKWBZFNQHAAO-UHFFFAOYSA-N
3-iodophenol
12272
220
99%

Chemical Identifiers

626-02-8
220
FXTKWBZFNQHAAO-UHFFFAOYSA-N
12272
3-iodophenol
C6H5IO
MFCD00002261
3-iodo-phenol, 3-jodphenol, 3-jodphenol german, 5-iodophenol, m-hydroxyiodobenzene, m-iodophenol, m-jodphenol, phenol, 3-iodo, phenol, m-iodo, pubchem3968
CHEBI:33439
C1=CC(=CC(=C1)I)O
Safety and Handling

Safety and Handling

GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 210-923-2

RTECSNumber : SL5490000

TSCA : TSCA

Recommended Storage : Product darkens during storage

SDS Documents