Learn More
3-Iodooxetane, 95%
CAS: 26272-85-5 | C3H5IO | 183.98 g/mol
Supplier: Thermo Scientific Chemicals H57302ME
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| 3-Iodooxetane | |
| 2.14 | |
| 54°C (129°F) | |
| MFCD08544403 | |
| UN1993 | |
| 3-iodo-oxetane, 1,3-epoxy-2-iodopropane, oxetane, 3-iodo, 3-iodanyloxetane, 3-iodooxetane, acmc-1cgai, 2-iodo-1,3-epoxypropane, 1,3-?epoxy-?2-?iodopropane | |
| IC1COC1 | |
| 183.98 | |
| 183.98 |
| 26272-85-5 | |
| 166°C | |
| C3H5IO | |
| 500 mg | |
| Light sensitive | |
| KBEIFKMKVCDETC-UHFFFAOYSA-N | |
| 3-iodooxetane | |
| 13356686 | |
| 95% |
Chemical Identifiers
| 26272-85-5 | |
| 183.98 | |
| KBEIFKMKVCDETC-UHFFFAOYSA-N | |
| 13356686 | |
| IC1COC1 |
| C3H5IO | |
| MFCD08544403 | |
| 3-iodo-oxetane, 1,3-epoxy-2-iodopropane, oxetane, 3-iodo, 3-iodanyloxetane, 3-iodooxetane, acmc-1cgai, 2-iodo-1,3-epoxypropane, 1,3-?epoxy-?2-?iodopropane | |
| 3-iodooxetane |
Safety and Handling
GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c
H226-H315-H319-H335
DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
TSCA : No
Recommended Storage : Keep cold
RUO – Research Use Only