missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-Iodocarbazole, TCI America™

$128.34 - $427.78

Chemical Identifiers

CAS 16807-13-9
Molecular Formula C12H8IN
Molecular Weight (g/mol) 293.107
MDL Number MFCD00267746
InChI Key OYIGWMXXIFYAGD-UHFFFAOYSA-N
PubChem CID 3678069
IUPAC Name 3-iodo-9H-carbazole
SMILES C1=CC=C2C(=C1)C3=C(N2)C=CC(=C3)I
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
I09195G View Documents TCI America
I09195G
5g Each for $128.34
 
I091925G View Documents TCI America
I091925G
25g Each for $427.78
 
Specifications

Chemical Identifiers

16807-13-9
293.107
OYIGWMXXIFYAGD-UHFFFAOYSA-N
3-iodo-9H-carbazole
C12H8IN
MFCD00267746
3678069
C1=CC=C2C(=C1)C3=C(N2)C=CC(=C3)I

Specifications

White
5g
C12H8IN
OYIGWMXXIFYAGD-UHFFFAOYSA-N
3-iodo-9H-carbazole
3678069
Crystal-Powder at 20°C
3-Iodocarbazole
199°C
16807-13-9
MFCD00267746
C1=CC=C2C(=C1)C3=C(N2)C=CC(=C3)I
293.107
293.11
≥98.0% (GC)
Safety and Handling

Safety and Handling

TSCA : No