Please login to your online account to display your discounted pricing

3-Iodobenzylamine Hydrochloride 97%, ACROS Organics™


Quantity 5g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  158970050

 View more versions of this product

Catalog No. AC158970050

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥96.0%
Physical Form Crystalline Powder
Formula Weight 269.51
Quantity 5g
Color Off-white to light beige
EINECS Number 223-068-5
TSCA TSCA
Melting Point 188° to 194°C
CAS Min % 96.0
CAS Max % 100.0
Chemical Name or Material 3-Iodobenzylamine hydrochloride
Infrared Spectrum Authentic
Packaging Glass bottle
Name Note 97%
Linear Formula IC6H4CH2NH2·HCl
CAS 3718-88-5
Assay Percent Range 97%
Molecular Formula C7H8IN·HCl
MDL Number MFCD00012857

Specifications

Chemical Name or Material 3-Iodobenzylamine hydrochloride
Name Note 97%
CAS 3718-88-5
CAS Min % 96.0
CAS Max % 100.0
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C7H8IN·HCl
Linear Formula IC6H4CH2NH2·HCl
Formula Weight 269.51
MDL Number MFCD00012857