Please login to your online account to display your discounted pricing

3-Iodobenzylamine Hydrochloride 97%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 3-Iodobenzylamine hydrochloride
Name Note 97%
CAS 3718-88-5
CAS Min % 96.0
CAS Max % 100.0
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C7H8IN·HCl
Linear Formula IC6H4CH2NH2·HCl
Formula Weight 269.51
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC158970050

  SDS
thermo fisher scientific
158970050
Glass bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥96.0%
Physical Form Crystalline Powder
Formula Weight 269.51
Quantity 5g
Color Off-white to light beige
EINECS Number 223-068-5
TSCA TSCA
Melting Point 188° to 194°C
CAS Min % 96.0
CAS Max % 100.0
Chemical Name or Material 3-Iodobenzylamine hydrochloride
Infrared Spectrum Authentic
Packaging Glass bottle
Name Note 97%
Linear Formula IC6H4CH2NH2·HCl
CAS 3718-88-5
Assay Percent Range 97%
Molecular Formula C7H8IN·HCl
MDL Number MFCD00012857