Please login to your online account to display your discounted pricing

3-Iodobenzylamine, 97%, ACROS Organics™


Quantity 20g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC367320200

 View more versions of this product

Catalog No. AC367320200

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥97.5%
Physical Form Liquid
Formula Weight 233.05
Quantity 20g
Color Clear light yellow
Density 1.7480g/mL
Beilstein 12, 2354
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 97%
Molecular Formula C7H8IN
CAS 696-40-2
Infrared Spectrum Authentic
Packaging Glass bottle
Refractive Index 1.6380 to 1.6400 (20°C, 589nm)
Linear Formula IC6H4CH2NH2
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
Health Hazard 3 GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Chemical Name or Material 3-Iodobenzylamine
MDL Number MFCD00192227

Specifications

Chemical Name or Material 3-Iodobenzylamine
CAS 696-40-2
CAS Min % 96.0
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 97%
Molecular Formula C7H8IN
Linear Formula IC6H4CH2NH2
Formula Weight 233.05
MDL Number MFCD00192227
Accessories

Danger

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.

SDS & Safety Documents