Please login to your online account to display your discounted pricing

3-Iodobenzylamine, 97%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 3-Iodobenzylamine
CAS 696-40-2
CAS Min % 96.0
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 97%
Molecular Formula C7H8IN
Linear Formula IC6H4CH2NH2
Formula Weight 233.05
MDL Number MFCD00192227
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC367320200

  SDS
thermo fisher scientific
AC367320200
Glass bottle 20g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC367320010

  SDS
thermo fisher scientific
AC367320010
Glass bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC367320050

  SDS
thermo fisher scientific
AC367320050
Glass bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC367322500

  SDS
thermo fisher scientific
AC367322500
Glass bottle 250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥97.5%
Physical Form Liquid
Formula Weight 233.05
Quantity 20g
Color Clear light yellow
Density 1.7480g/mL
Beilstein 12, 2354
CAS Min % 96.0
CAS Max % 100.0
Molecular Formula C7H8IN
CAS 696-40-2
Assay Percent Range 97%
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
Health Hazard 3 GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Packaging Glass bottle
Refractive Index 1.6380 to 1.6400 (20°C, 589nm)
Infrared Spectrum Authentic
Linear Formula IC6H4CH2NH2
Chemical Name or Material 3-Iodobenzylamine
MDL Number MFCD00192227
Safety and Handling

Danger

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.