missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-Iodobenzylamine, 97%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals L1780106

 View more versions of this product

Catalog No. AAL1780106


Description

Description

It is a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is a pharmaceutical intermediate.

Solubility
Difficult to mix in water.

Notes
Air sensitive and light sensitive. Store in cool dry conditions in well sealed containers. Incompatible with strong oxidizing agents.
Specifications

Specifications

3-Iodobenzylamine
1.748
>110°C (230°F)
1.6375
5 g
2689602
3-iodobenzylamine, 3-iodophenyl methanamine, meta-iodobenzylamine, benzenemethanamine, 3-iodo, 3-iodo-benzylamine, 1-3-iodophenyl methanamine, 3-iodobenzyl amine, m-iodobenzylamine, 3-iodobenzenemethanamine, 3-iodo-benzenemethanamine
LQLOGZQVKUNBRX-UHFFFAOYSA-N
(3-iodophenyl)methanamine
97012
97%
696-40-2
132°C (8 mmHg)
C7H8IN
MFCD00192227
UN2735
Air and light sensitive
Difficult to mix in water.
C1=CC(=CC(=C1)I)CN
233.052
233.05

Chemical Identifiers

696-40-2
233.052
LQLOGZQVKUNBRX-UHFFFAOYSA-N
97012
C1=CC(=CC(=C1)I)CN
C7H8IN
MFCD00192227
3-iodobenzylamine, 3-iodophenyl methanamine, meta-iodobenzylamine, benzenemethanamine, 3-iodo, 3-iodo-benzylamine, 1-3-iodophenyl methanamine, 3-iodobenzyl amine, m-iodobenzylamine, 3-iodobenzenemethanamine, 3-iodo-benzenemethanamine
(3-iodophenyl)methanamine
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c

H314

DOTInformation : Hazard Class: 8; Packaging Group: III

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only