missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Iodobenzyl bromide, 96%

Manufacturer:  Alfa Aesar™ A1114022

 View more versions of this product

Catalog No. AAA1114022



Specifications

Specifications

3-Iodobenzyl bromide
106°C (1mmHg)
100g
C7H6BrI
UN3261
3-iodobenzyl bromide, 1-bromomethyl-3-iodobenzene, alpha-bromo-3-iodotoluene, 3-iodobenzylbromide, m-iodobenzyl bromide, benzene, 1-bromomethyl-3-iodo, m-iodobenzylbromide, 3-iodo-benzylbromide, 3-iodo-benzyl bromide, meta-iodobenzyl bromide
C1=CC(=CC(=C1)I)CBr
296.933
296.94
>110°C (230°F)
48°C to 51°C
49617-83-6
MFCD00019016
742268
BACZSVQZBSCWIG-UHFFFAOYSA-N
1-(bromomethyl)-3-iodobenzene
2759361
96%

Chemical Identifiers

49617-83-6
296.933
BACZSVQZBSCWIG-UHFFFAOYSA-N
2759361
C1=CC(=CC(=C1)I)CBr
C7H6BrI
MFCD00019016
3-iodobenzyl bromide, 1-bromomethyl-3-iodobenzene, alpha-bromo-3-iodotoluene, 3-iodobenzylbromide, m-iodobenzyl bromide, benzene, 1-bromomethyl-3-iodo, m-iodobenzylbromide, 3-iodo-benzylbromide, 3-iodo-benzyl bromide, meta-iodobenzyl bromide
1-(bromomethyl)-3-iodobenzene
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.

SDS Documents