Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Iodobenzyl alcohol, 98%

$28.32 - $86.50

Chemical Identifiers

Chemical Name or Material 3-Iodobenzyl alcohol
CAS 57455-06-8
Molecular Formula C7H7IO
Formula Weight 234.04
MDL Number MFCD00004635
Beilstein 3234821
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0530903

  SDS
alfa aesar™
L0530903
1g Each for $28.32

AAL0530906

  SDS
alfa aesar™
L0530906
5g Each for $86.50
Description & Specifications

Specifications

Formula Weight 234.04
Chemical Name or Material 3-Iodobenzyl alcohol
Molecular Formula C7H7IO
MDL Number MFCD00004635
CAS 57455-06-8
Assay 98%
Health Hazard 1 Warning
Health Hazard 3 GHS P Statement
P280-P305+P351+P338-P362-P321-P332+P313-P337+P313
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Take off contaminated clothing and wash before reuse.
Specific treatment (see label).
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.
Quantity 1g
Health Hazard 2 GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
Beilstein 3234821
Boiling Point 254° to 255°C
Flash Point >110°C (230°F)
RTECS Number DO8318000
Safety and Handling

Warning

GHS P Statement
P280-P305+P351+P338-P362-P321-P332+P313-P337+P313
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Take off contaminated clothing and wash before reuse.
Specific treatment (see label).
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.

GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.