missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-Iodobenzoyl Chloride, TCI America™

$55.00 - $168.06

Chemical Identifiers

CAS 1711-10-0
Molecular Formula C7H4ClIO
Molecular Weight (g/mol) 266.462
MDL Number MFCD02093934
InChI Key DTELTOREECFDBC-UHFFFAOYSA-N
Synonym 3-iodobenzoylchloride, m-iodobenzoyl chloride, benzoyl chloride, 3-iodo, 3-iodo benzoyl chloride, 3-iodo-benzoyl chloride, benzoylchloride,3-iodo, benzoyl chloride,3-iodo, acmc-1bql4, 3-iodobenzoic acid chloride, 3-iodobenzoyl chloride, 97+%
PubChem CID 74378
IUPAC Name 3-iodobenzoyl chloride
SMILES C1=CC(=CC(=C1)I)C(=O)Cl
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
I07205G View Documents TCI America
I07205G
5g Each for $168.06
 
I07201G View Documents TCI America
I07201G
1g Each for $55.00
 
Specifications

Chemical Identifiers

1711-10-0
266.462
DTELTOREECFDBC-UHFFFAOYSA-N
74378
C1=CC(=CC(=C1)I)C(=O)Cl
C7H4ClIO
MFCD02093934
3-iodobenzoylchloride, m-iodobenzoyl chloride, benzoyl chloride, 3-iodo, 3-iodo benzoyl chloride, 3-iodo-benzoyl chloride, benzoylchloride,3-iodo, benzoyl chloride,3-iodo, acmc-1bql4, 3-iodobenzoic acid chloride, 3-iodobenzoyl chloride, 97+%
3-iodobenzoyl chloride

Specifications

146°C
25°C
1711-10-0
MFCD02093934
3-iodobenzoylchloride, m-iodobenzoyl chloride, benzoyl chloride, 3-iodo, 3-iodo benzoyl chloride, 3-iodo-benzoyl chloride, benzoylchloride,3-iodo, benzoyl chloride,3-iodo, acmc-1bql4, 3-iodobenzoic acid chloride, 3-iodobenzoyl chloride, 97+%
C1=CC(=CC(=C1)I)C(=O)Cl
266.462
266.46
≥98.0% (GC,T)
Undesignated
5g
C7H4ClIO
3261
DTELTOREECFDBC-UHFFFAOYSA-N
3-iodobenzoyl chloride
74378
Crystal at 20°C
3-Iodobenzoyl Chloride
Safety and Handling

Safety and Handling

TSCA : No