missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Iodobenzoyl chloride, 96%

$64.77 - $64.77

Chemical Identifiers

CAS 1711-10-0
Molecular Formula C7H4ClIO
Molecular Weight (g/mol) 266.462
MDL Number MFCD02093934
InChI Key DTELTOREECFDBC-UHFFFAOYSA-N
Synonym 3-iodobenzoylchloride, m-iodobenzoyl chloride, benzoyl chloride, 3-iodo, 3-iodo benzoyl chloride, 3-iodo-benzoyl chloride, benzoylchloride,3-iodo, benzoyl chloride,3-iodo, acmc-1bql4, 3-iodobenzoic acid chloride, 3-iodobenzoyl chloride, 97+%
PubChem CID 74378
IUPAC Name 3-iodobenzoyl chloride
SMILES C1=CC(=CC(=C1)I)C(=O)Cl
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL1745103 View Documents Alfa Aesar™
L1745103
1g Each for $64.77
 
Description

Description

Applications
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs.

Solubility
Reacts with water.

Notes
Moisture and light sensitive. Store in dry condition. Keep away from active metals, bases, oxidizing agents and heat.
Specifications

Chemical Identifiers

1711-10-0
266.462
DTELTOREECFDBC-UHFFFAOYSA-N
74378
C1=CC(=CC(=C1)I)C(=O)Cl
C7H4ClIO
MFCD02093934
3-iodobenzoylchloride, m-iodobenzoyl chloride, benzoyl chloride, 3-iodo, 3-iodo benzoyl chloride, 3-iodo-benzoyl chloride, benzoylchloride,3-iodo, benzoyl chloride,3-iodo, acmc-1bql4, 3-iodobenzoic acid chloride, 3-iodobenzoyl chloride, 97+%
3-iodobenzoyl chloride

Specifications

159°C to 160°C (23mmHg)
1g
C7H4ClIO
UN3261
3-iodobenzoylchloride, m-iodobenzoyl chloride, benzoyl chloride, 3-iodo, 3-iodo benzoyl chloride, 3-iodo-benzoyl chloride, benzoylchloride,3-iodo, benzoyl chloride,3-iodo, acmc-1bql4, 3-iodobenzoic acid chloride, 3-iodobenzoyl chloride, 97+%
C1=CC(=CC(=C1)I)C(=O)Cl
266.462
266.47
3-Iodobenzoyl chloride
23°C to 25°C
1711-10-0
MFCD02093934
1860418
DTELTOREECFDBC-UHFFFAOYSA-N
3-iodobenzoyl chloride
74378
96%
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement
P280-P305+P351+P338-P309-P310
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.

Danger

DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.