Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Iodobenzonitrile, 99%

Chemical Identifiers

Chemical Name or Material 3-Iodobenzonitrile
CAS 69113-59-3
Molecular Formula C7H4IN
Formula Weight 229.02
MDL Number MFCD00079762
UN Number UN3439
Synonym 3-Cyanoiodobenzene
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2202009

  SDS
alfa aesar™
B2202009
10g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAB2202088

  SDS
alfa aesar™
B2202088
2.5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 229.02
Melting Point 38° to 42°C
Quantity 10g
Health Hazard 1 Danger
MDL Number MFCD00079762
DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.
Health Hazard 3 GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
CAS 69113-59-3
Chemical Name or Material 3-Iodobenzonitrile
Assay 99%
Health Hazard 2 GHS H Statement
H301-H312-H332-H315-H319-H335
Toxic if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
UN Number UN3439
Synonym 3-Cyanoiodobenzene
Molecular Formula C7H4IN
Flash Point >110°C (230°F)
Safety and Handling

Danger

GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301-H312-H332-H315-H319-H335
Toxic if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.