missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
3-Iodoanisole, 97%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 222020050
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
3-Iodoanisole | |
96.0 | |
1.9600g/mL | |
>110°C | |
96% min. (GC) | |
C7H7IO | |
IC6H4OCH3 | |
5g | |
1.96 | |
RSHBAGGASAJQCH-UHFFFAOYSA-N | |
1-iodo-3-methoxybenzene | |
69839 | |
97% |
766-85-8 | |
100.0 | |
244°C to 245°C | |
Authentic | |
Glass bottle | |
1.6130 to 1.615 | |
MFCD00001048 | |
06, 208 | |
3-iodoanisole, m-iodoanisole, benzene, 1-iodo-3-methoxy, anisole, m-iodo, m-methoxyiodobenzene, 3-methoxyphenyl iodide, 3-methoxyiodobenzene, 1-iodo-3-methoxy-benzene, m-iodoanisol, m-iodo-anisole | |
COC1=CC(=CC=C1)I | |
234.04 | |
234.04 |
Chemical Identifiers
766-85-8 | |
234.04 | |
RSHBAGGASAJQCH-UHFFFAOYSA-N | |
69839 | |
COC1=CC(=CC=C1)I |
C7H7IO | |
MFCD00001048 | |
3-iodoanisole, m-iodoanisole, benzene, 1-iodo-3-methoxy, anisole, m-iodo, m-methoxyiodobenzene, 3-methoxyphenyl iodide, 3-methoxyiodobenzene, 1-iodo-3-methoxy-benzene, m-iodoanisol, m-iodo-anisole | |
1-iodo-3-methoxybenzene |
Safety and Handling
EINECSNumber : 212-173-1
Recommended Storage : Product may on darken on storage
RUO â Research Use Only