Learn More
3-Iodoaniline, 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals L0115909
Description
3-Iodoanilineis used as Anilines, Aromatic Amines and Nitro Compounds.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications3-Iodoanilineis used as Anilines, Aromatic Amines and Nitro Compounds.
Solubility
Insoluble in water.
Notes
Light Sensitive. Store at 4°C. Store protected from light. Incompatible with Acid chlorides, acid anhydrides, acids, chloroformates, strong oxidizing agents.
Specifications
3-Iodoaniline | |
21°C to 24°C | |
145°C to 146°C (15 mmHg) | |
C6H6IN | |
MFCD00007781 | |
636058 | |
m-iodoaniline, benzenamine, 3-iodo, aniline, m-iodo, m-aminoiodobenzene, 3-iodobenzenamine, 1-amino-3-iodobenzene, 3-iodo-phenylamine, 3iodoaniline, 3-iodo-aniline, 3-iodophenylamine | |
FFCSRWGYGMRBGD-UHFFFAOYSA-N | |
3-iodoaniline | |
12271 | |
98% |
626-01-7 | |
1.821 | |
>110°C (230°F) | |
1.684 | |
10 g | |
Light sensitive | |
Insoluble in water. | |
C1=CC(=CC(=C1)I)N | |
219.025 | |
219.03 |
Chemical Identifiers
626-01-7 | |
219.025 | |
FFCSRWGYGMRBGD-UHFFFAOYSA-N | |
12271 | |
C1=CC(=CC(=C1)I)N |
C6H6IN | |
MFCD00007781 | |
m-iodoaniline, benzenamine, 3-iodo, aniline, m-iodo, m-aminoiodobenzene, 3-iodobenzenamine, 1-amino-3-iodobenzene, 3-iodo-phenylamine, 3iodoaniline, 3-iodo-aniline, 3-iodophenylamine | |
3-iodoaniline |
Safety and Handling
GHS H Statement
H302-H312-H332-H315-H319
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302+H312+H332-H315-H319-H335
DOTInformation : Hazard Class: 6.1; Packaging Group: III
EINECSNumber : 210-922-7
RTECSNumber : BY3750000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only