missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-Iodo-o-xylene, TCI America™

$143.61 - $424.73

Chemical Identifiers

CAS 31599-60-7
Molecular Formula C8H9I
Molecular Weight (g/mol) 232.064
MDL Number MFCD00041437
InChI Key DANMWBNOPFBJSZ-UHFFFAOYSA-N
Synonym 3-iodo-o-xylene, 1,2-dimethyl-3-iodobenzene, 2,3-dimethyliodobenzene, benzene, 1-iodo-2,3-dimethyl, benzene, iododimethyl, 2,3-dimethyl-1-iodobenzene, jodxylol, 3-iodo-ortho-xylene, pubchem3865, acmc-1ajf2
PubChem CID 141645
IUPAC Name 1-iodo-2,3-dimethylbenzene
SMILES CC1=C(C(=CC=C1)I)C
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
I054725G View Documents TCI America
I054725G
25g Each for $424.73
 
I05475G View Documents TCI America
I05475G
5g Each for $143.61
 
Specifications

Chemical Identifiers

31599-60-7
232.064
DANMWBNOPFBJSZ-UHFFFAOYSA-N
141645
CC1=C(C(=CC=C1)I)C
C8H9I
MFCD00041437
3-iodo-o-xylene, 1,2-dimethyl-3-iodobenzene, 2,3-dimethyliodobenzene, benzene, 1-iodo-2,3-dimethyl, benzene, iododimethyl, 2,3-dimethyl-1-iodobenzene, jodxylol, 3-iodo-ortho-xylene, pubchem3865, acmc-1ajf2
1-iodo-2,3-dimethylbenzene

Specifications

231°C
25g
C8H9I
3-iodo-o-xylene, 1,2-dimethyl-3-iodobenzene, 2,3-dimethyliodobenzene, benzene, 1-iodo-2,3-dimethyl, benzene, iododimethyl, 2,3-dimethyl-1-iodobenzene, jodxylol, 3-iodo-ortho-xylene, pubchem3865, acmc-1ajf2
CC1=C(C(=CC=C1)I)C
232.064
232.06
≥97.0% (GC)
Undesignated
31599-60-7
MFCD00041437
DANMWBNOPFBJSZ-UHFFFAOYSA-N
1-iodo-2,3-dimethylbenzene
141645
Clear Liquid at 20°C
3-Iodo-o-xylene
Safety and Handling

Safety and Handling

TSCA : No