missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-Iodo-6-nitroindazole, TCI America™

$145.14 - $433.89

Chemical Identifiers

CAS 70315-70-7
Molecular Formula C7H4IN3O2
Molecular Weight (g/mol) 289.032
MDL Number MFCD07781652
InChI Key GZCGNGLOCQEDMT-UHFFFAOYSA-N
PubChem CID 12475306
IUPAC Name 3-iodo-6-nitro-2H-indazole
SMILES C1=CC2=C(NN=C2C=C1[N+](=O)[O-])I
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
I10041G View Documents TCI America
I10041G
1g Each for $145.14
 
I10045G View Documents TCI America
I10045G
5g Each for $433.89
 
Specifications

Chemical Identifiers

70315-70-7
289.032
GZCGNGLOCQEDMT-UHFFFAOYSA-N
3-iodo-6-nitro-2H-indazole
C7H4IN3O2
MFCD07781652
12475306
C1=CC2=C(NN=C2C=C1[N+](=O)[O-])I

Specifications

Undesignated
1g
C7H4IN3O2
GZCGNGLOCQEDMT-UHFFFAOYSA-N
3-iodo-6-nitro-2H-indazole
12475306
Crystal-Powder at 20°C
3-Iodo-6-nitroindazole
265°C
70315-70-7
MFCD07781652
C1=CC2=C(NN=C2C=C1[N+](=O)[O-])I
289.032
289.03
≥98.0% (GC)
Safety and Handling

Safety and Handling

TSCA : No