Please login to your online account to display your discounted pricing

3-Iodo-4-nitrotoluene, 97%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 3-Iodo-4-nitrotoluene
CAS 52488-29-6
CAS Min % 96.0
CAS Max % 100.0
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C7H6INO2
Formula Weight 263.03
MDL Number MFCD01320681
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC326290050

  SDS
thermo fisher scientific
AC326290050
Glass bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥96.0%
Physical Form Low Melting Crystalline Mass or Liquid
Formula Weight 263.03
Quantity 5g
Color Yellow
Beilstein 05, II, 258
CAS Max % 100.0
CAS Min % 96.0
CAS 52488-29-6
Chemical Name or Material 3-Iodo-4-nitrotoluene
Molecular Formula C7H6INO2
Assay Percent Range 97%
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Infrared Spectrum Authentic
MDL Number MFCD01320681
Packaging Glass bottle
Health Hazard 2 GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.