Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Iodo-4-methylbenzoic acid, 98%

$49.11 - $568.05

Chemical Identifiers

Chemical Name or Material 3-Iodo-4-methylbenzoic acid
CAS 82998-57-0
Molecular Formula C8H7IO2
Formula Weight 262.05
MDL Number MFCD00010392
Beilstein 2436216
Synonym 3-Iodo-p-toluic acid
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1419706

 
alfa aesar™
A1419706
5g Each for $49.11

AAA1419714

 
alfa aesar™
A1419714
25g Each for $171.77

AAA1419722

 
alfa aesar™
A1419722
100g Each for $568.05
Description & Specifications

Specifications

Formula Weight 262.05
Synonym 3-Iodo-p-toluic acid
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Molecular Formula C8H7IO2
Melting Point 206° to 210°C
CAS 82998-57-0
Beilstein 2436216
Health Hazard 1 Warning
Quantity 5g
Chemical Name or Material 3-Iodo-4-methylbenzoic acid
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Assay 98%
MDL Number MFCD00010392

Applications
3-Iodo-4-methylbenzoic acid has been used in the preparation of unlabeled N-succinimidyl 4-guanidinomethyl-3-iodobenzoate (SGIMB) and boc-protected derivative of SGIMB (Boc-SGMIB).

Solubility
Soluble in methanol.

Notes
Recommended storage temp 2-8°C. Store in cool, well ventilated area. Keep container tightly closed. Light Sensitive.

Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.