missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Iodo-4-methoxybenzaldehyde, 96%

$83.58 - $293.00

Chemical Identifiers

CAS 2314-37-6
Molecular Formula C8H7IO2
Molecular Weight (g/mol) 262.046
MDL Number MFCD01009500
InChI Key RVWOHBWQJGLXIJ-UHFFFAOYSA-N
Synonym 3-iodo-4-methoxy-benzaldehyde, benzaldehyde, 3-iodo-4-methoxy, 3-iodo-p-anisaldehyde, 4-formyl-2-iodoanisole, acmc-1cpd6, 3-iodanyl-4-methoxy-benzaldehyde, 3-iodo-4-methoxybenzaldehyde, benzaldehyde,3-iodo-4-methoxy
PubChem CID 1494376
IUPAC Name 3-iodo-4-methoxybenzaldehyde
SMILES COC1=C(C=C(C=C1)C=O)I
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH6622203 View Documents Alfa Aesar™
H6622203
1g Each for $83.58
 
AAH6622206 View Documents Alfa Aesar™
H6622206
5g Each for $293.00
 
Specifications

Chemical Identifiers

2314-37-6
262.046
RVWOHBWQJGLXIJ-UHFFFAOYSA-N
1494376
COC1=C(C=C(C=C1)C=O)I
C8H7IO2
MFCD01009500
3-iodo-4-methoxy-benzaldehyde, benzaldehyde, 3-iodo-4-methoxy, 3-iodo-p-anisaldehyde, 4-formyl-2-iodoanisole, acmc-1cpd6, 3-iodanyl-4-methoxy-benzaldehyde, 3-iodo-4-methoxybenzaldehyde, benzaldehyde,3-iodo-4-methoxy
3-iodo-4-methoxybenzaldehyde

Specifications

Air & Light Sensitive
107°C to 112°C
2314-37-6
MFCD01009500
RVWOHBWQJGLXIJ-UHFFFAOYSA-N
3-iodo-4-methoxybenzaldehyde
1494376
Solid
3-Iodo-4-methoxybenzaldehyde
White to White-Yellow
1g
C8H7IO2
3-iodo-4-methoxy-benzaldehyde, benzaldehyde, 3-iodo-4-methoxy, 3-iodo-p-anisaldehyde, 4-formyl-2-iodoanisole, acmc-1cpd6, 3-iodanyl-4-methoxy-benzaldehyde, 3-iodo-4-methoxybenzaldehyde, benzaldehyde,3-iodo-4-methoxy
COC1=C(C=C(C=C1)C=O)I
262.046
262.04
96%