Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Iodo-4,5-dimethoxybenzaldehyde, 98%

$96.97 - $96.97

Chemical Identifiers

Chemical Name or Material 3-Iodo-4,5-dimethoxybenzaldehyde
CAS 32024-15-0
Molecular Formula C9H9IO3
Formula Weight 292.07
MDL Number MFCD00674091
Beilstein 1961757
Synonym 4,5-Dimethoxy-3-iodobenzaldehyde; 5-Iodoveratraldehyde
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1745206

 
alfa aesar™
L1745206
5g Each for $96.97
Description & Specifications

Specifications

Formula Weight 292.07
Chemical Name or Material 3-Iodo-4,5-dimethoxybenzaldehyde
Assay 98%
Molecular Formula C9H9IO3
MDL Number MFCD00674091
CAS 32024-15-0
Synonym 4,5-Dimethoxy-3-iodobenzaldehyde; 5-Iodoveratraldehyde
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Melting Point 68° to 70°C
Quantity 5g
Beilstein 1961757
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.