missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Iodo-4,5-dimethoxybenzaldehyde, 98%

Manufacturer:  Alfa Aesar™ L1745203

 View more versions of this product

Catalog No. AAL1745203



Specifications

Specifications

3-Iodo-4,5-dimethoxybenzaldehyde
1g
C9H9IO3
1961757
MVPNBXPAUYYZAF-UHFFFAOYSA-N
3-iodo-4,5-dimethoxybenzaldehyde
2778113
98%
68°C to 70°C
32024-15-0
MFCD00674091
3,4-dimethoxy-5-iodo-benzaldehyde
COC1=C(C(=CC(=C1)C=O)I)OC
292.072
292.07

Chemical Identifiers

32024-15-0
292.072
MVPNBXPAUYYZAF-UHFFFAOYSA-N
2778113
COC1=C(C(=CC(=C1)C=O)I)OC
C9H9IO3
MFCD00674091
3,4-dimethoxy-5-iodo-benzaldehyde
3-iodo-4,5-dimethoxybenzaldehyde
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

Documents