Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Iodo-4-(2,2,2-trimethylacetamido)pyridine

$242.59 - $1,074.32

Chemical Identifiers

Chemical Name or Material 3-Iodo-4-(2,2,2-trimethylacetamido)pyridine
CAS 113975-33-0
Molecular Formula C10H13IN2O
Formula Weight 304.13
MDL Number MFCD04971323
Synonym N-(3-Iodo-4-pyridyl)pivalamide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH5002603

  SDS
alfa aesar™
H5002603
1g Each for $242.59

AAH5002606

  SDS
alfa aesar™
H5002606
5g Each for $1,074.32
Description & Specifications

Specifications

Formula Weight 304.13
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Health Hazard 1 Warning
CAS 113975-33-0
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Chemical Name or Material 3-Iodo-4-(2,2,2-trimethylacetamido)pyridine
Synonym N-(3-Iodo-4-pyridyl)pivalamide
MDL Number MFCD04971323
Quantity 1g
Molecular Formula C10H13IN2O
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.