Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Iodo-2-isopropoxypyridine, 99%

$204.92 - $907.07

Chemical Identifiers

Chemical Name or Material 3-Iodo-2-isopropoxypyridine
CAS 766557-61-3
Molecular Formula C8H10INO
Formula Weight 263.08
MDL Number MFCD06659009
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH5008303

  SDS
alfa aesar™
H5008303
1g Each for $204.92

AAH5008306

  SDS
alfa aesar™
H5008306
5g Each for $907.07
Description & Specifications

Specifications

Formula Weight 263.08
Chemical Name or Material 3-Iodo-2-isopropoxypyridine
Assay 99%
Molecular Formula C8H10INO
MDL Number MFCD06659009
CAS 766557-61-3
Quantity 1g
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.