missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Iodo-2-(2,2,2-trifluoroethoxy)pyridine

$1,066.59 - $1,066.59

Chemical Identifiers

CAS 912761-82-1
Molecular Formula C7H5F3INO
Formula Weight 303.02
MDL Number MFCD07781168
Chemical Name or Material 3-Iodo-2-(2,2,2-trifluoroethoxy)pyridine
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH5010806 SDS Alfa Aesar™
H5010806
5g Each for $1,066.59
AAH5010803 SDS Alfa Aesar™
H5010803
1g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

912761-82-1
303.02
3-Iodo-2-(2,2,2-trifluoroethoxy)pyridine
C7H5F3INO
MFCD07781168

Specifications

912761-82-1
303.02
5g
C7H5F3INO
MFCD07781168
3-Iodo-2-(2,2,2-trifluoroethoxy)pyridine

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning