missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Iodo-1,2,4,5-tetramethylbenzene, 98%

$37.39 - $146.59

Chemical Identifiers

CAS 2100-25-6
Molecular Formula C10H13I
Formula Weight 260.12
MDL Number MFCD00013705
Beilstein 1862299
Synonym 2,3,5,6-Tetramethyliodobenzene
Chemical Name or Material 3-Iodo-1,2,4,5-tetramethylbenzene
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA1571614 SDS Alfa Aesar™
A1571614
25g Each for $146.59
AAA1571606 SDS Alfa Aesar™
A1571606
5g Each for $37.39
Specifications Description & Specifications

Chemical Identifiers

2100-25-6
260.12
1862299
3-Iodo-1,2,4,5-tetramethylbenzene
C10H13I
MFCD00013705
2,3,5,6-Tetramethyliodobenzene

Specifications

2100-25-6
260.12
1862299
2,3,5,6-Tetramethyliodobenzene
3-Iodo-1,2,4,5-tetramethylbenzene
C10H13I
MFCD00013705
78° to 80°C
25g
98%