missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Alfa Aesar™ 3-Iodo-1,2,4,5-tetramethylbenzene, 98%
Manufacturer: Alfa Aesar™ A1571614
Specifications
3-Iodo-1,2,4,5-tetramethylbenzene | |
25g | |
C10H13I | |
1862299 | |
MCRSYYLHVQGFNR-UHFFFAOYSA-N | |
3-iodo-1,2,4,5-tetramethylbenzene | |
75011 | |
98% |
78°C to 80°C | |
2100-25-6 | |
MFCD00013705 | |
2,3,5,6-tetramethyliodobenzene, iododurene, 3-iododurene, benzene, 3-iodo-1,2,4,5-tetramethyl, 1-iodo-2,3,5,6-tetramethylbenzene, 3-iodo-1,2,4,5-tetramethyl-benzene, acmc-209fgl, maybridge1_002808, 3-iodo-1,4,5-tetramethylbenzene, iodo-2,3,5,6-tetramethylbenzene | |
CC1=CC(=C(C(=C1C)I)C)C | |
260.118 | |
260.12 |
Chemical Identifiers
2100-25-6 | |
260.118 | |
MCRSYYLHVQGFNR-UHFFFAOYSA-N | |
75011 | |
CC1=CC(=C(C(=C1C)I)C)C |
C10H13I | |
MFCD00013705 | |
2,3,5,6-tetramethyliodobenzene, iododurene, 3-iododurene, benzene, 3-iodo-1,2,4,5-tetramethyl, 1-iodo-2,3,5,6-tetramethylbenzene, 3-iodo-1,2,4,5-tetramethyl-benzene, acmc-209fgl, maybridge1_002808, 3-iodo-1,4,5-tetramethylbenzene, iodo-2,3,5,6-tetramethylbenzene | |
3-iodo-1,2,4,5-tetramethylbenzene |