Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Indoxyl acetate, 97%

$65.09 - $65.09

Chemical Identifiers

Chemical Name or Material 3-Indoxyl acetate
CAS 608-08-2
Molecular Formula C10H9NO2
Formula Weight 175.19
MDL Number MFCD00014561
Beilstein 143086
Synonym 3-Acetoxyindole; Indolyl acetate
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0493203

  SDS
alfa aesar™
L0493203
1g Each for $65.09
Description & Specifications

Specifications

Formula Weight 175.19
MDL Number MFCD00014561
Assay 97%
Synonym 3-Acetoxyindole; Indolyl acetate
Chemical Name or Material 3-Indoxyl acetate
Molecular Formula C10H9NO2
Health Hazard 1 Warning
CAS 608-08-2
Quantity 1g
Health Hazard 2 GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Beilstein 143086
RTECS Number AI3325000
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
TSCA TSCA
Melting Point 128° to 130°C

Applications
Useful for detection of esterases3-Indoxyl acetate acts as a colorimetric substrate used for differentiating esterases from various microbiological species. Further, it is used as a plant growth-stimulating hormone and reagent. In addition to this, it acts as a useful synthetic intermediate.

Solubility
Soluble in ethanol, water, chloroform, dichloromethane and methanol.

Notes
Light sensitive. Store in a cool place. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with oxidizing agents.

Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.