Please login to your online account to display your discounted pricing

3-Indolepropionic acid, 99%, ACROS Organics™


Quantity 1g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC204990010

 View more versions of this product

Catalog No. AC204990010

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Physical Form Crystalline Powder
Assay ≥98.5% (HPLC)
Formula Weight 189.21
Quantity 1g
Color Yellow
Beilstein 22, 69
Water 0.5% Max. (K.F.)
Melting Point 130° to 136°C
CAS Max % 100.0
Chemical Name or Material 3-Indolepropionic acid
Infrared Spectrum Authentic
Assay Percent Range 99%
Molecular Formula C11H11NO2
CAS 830-96-6
MDL Number MFCD00005660
CAS Min % 98.5
Health Hazard 1 Warning
Packaging Glass bottle
Synonym 3-(1H-indol-3-yl)propanoic acid
Health Hazard 2 GHS H Statement:
Causes skin irritation.
Harmful if swallowed.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Specifications

Chemical Name or Material 3-Indolepropionic acid
CAS 830-96-6
CAS Min % 98.5
CAS Max % 100.0
Assay ≥98.5% (HPLC)
Assay Percent Range 99%
Molecular Formula C11H11NO2
Formula Weight 189.21
MDL Number MFCD00005660
Synonym 3-(1H-indol-3-yl)propanoic acid
Accessories

Warning

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
Harmful if swallowed.
Causes serious eye irritation.
May cause respiratory irritation.

SDS & Safety Documents