missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-Indoleacetonitrile 98.0+%, TCI America™

$70.77 - $474.02

Chemical Identifiers

CAS 771-51-7
Molecular Formula C10H8N2
Molecular Weight (g/mol) 156.188
MDL Number MFCD00005628
InChI Key DMCPFOBLJMLSNX-UHFFFAOYSA-N
Synonym 1h-indole-3-acetonitrile, 2-1h-indol-3-yl acetonitrile, 3-cyanomethyl indole, 3-indolacetonitrile, 3-indoleacetonitrile, 3-indolylacetonitrile, indole-3-acetonitrile, indoleacetonitrile, indolylacetonitril, indolylacetonitrile
PubChem CID 351795
ChEBI CHEBI:17566
IUPAC Name 2-(1H-indol-3-yl)acetonitrile
SMILES C1=CC=C2C(=C1)C(=CN2)CC#N
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
I00241G View Documents Tci America
I00241G
1g Each for $70.77
 
I002425G View Documents Tci America
I002425G
25g Each for $474.02
 
Specifications

Chemical Identifiers

771-51-7
156.188
DMCPFOBLJMLSNX-UHFFFAOYSA-N
351795
2-(1H-indol-3-yl)acetonitrile
C10H8N2
MFCD00005628
1h-indole-3-acetonitrile, 2-1h-indol-3-yl acetonitrile, 3-cyanomethyl indole, 3-indolacetonitrile, 3-indoleacetonitrile, 3-indolylacetonitrile, indole-3-acetonitrile, indoleacetonitrile, indolylacetonitril, indolylacetonitrile
CHEBI:17566
C1=CC=C2C(=C1)C(=CN2)CC#N

Specifications

Undesignated
1g
C10H8N2
3439
DMCPFOBLJMLSNX-UHFFFAOYSA-N
2-(1H-indol-3-yl)acetonitrile
351795
156.19
≥98.0% (GC)
37°C
771-51-7
MFCD00005628
1h-indole-3-acetonitrile, 2-1h-indol-3-yl acetonitrile, 3-cyanomethyl indole, 3-indolacetonitrile, 3-indoleacetonitrile, 3-indolylacetonitrile, indole-3-acetonitrile, indoleacetonitrile, indolylacetonitril, indolylacetonitrile
C1=CC=C2C(=C1)C(=CN2)CC#N
156.188
CHEBI:17566
Crystal-Powder at 20°C
3-Indoleacetonitrile
Safety and Handling

Safety and Handling

RTECSNumber : AM0700000

TSCA : Yes

Recommended Storage : Refrigerator