missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-Indazolinone, TCI America™

$126.81 - $441.53

Chemical Identifiers

CAS 7364-25-2
Molecular Formula C7H6N2O
Molecular Weight (g/mol) 134.138
MDL Number MFCD00005685
InChI Key SWEICGMKXPNXNU-UHFFFAOYSA-N
Synonym 1h-indazol-3-ol, 3-indazolinone, indazolinone, 1h-indazol-3 2h-one, 3-hydroxy-1h-indazole, 3-hydroxyindazole, 1,2-dihydro-3h-indazol-3-one, 3-oxo-1,2-indazole, 3h-indazol-3-one, 1,2-dihydro, 2h-indazol-3-ol
PubChem CID 81829
IUPAC Name 1,2-dihydroindazol-3-one
SMILES C1=CC=C2C(=C1)C(=O)NN2
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
I09291G View Documents TCI America
I09291G
1g Each for $126.81
 
I09295G View Documents TCI America
I09295G
5g Each for $441.53
 
Specifications

Chemical Identifiers

7364-25-2
134.138
SWEICGMKXPNXNU-UHFFFAOYSA-N
81829
C1=CC=C2C(=C1)C(=O)NN2
C7H6N2O
MFCD00005685
1h-indazol-3-ol, 3-indazolinone, indazolinone, 1h-indazol-3 2h-one, 3-hydroxy-1h-indazole, 3-hydroxyindazole, 1,2-dihydro-3h-indazol-3-one, 3-oxo-1,2-indazole, 3h-indazol-3-one, 1,2-dihydro, 2h-indazol-3-ol
1,2-dihydroindazol-3-one

Specifications

Undesignated
1g
C7H6N2O
1h-indazol-3-ol, 3-indazolinone, indazolinone, 1h-indazol-3 2h-one, 3-hydroxy-1h-indazole, 3-hydroxyindazole, 1,2-dihydro-3h-indazol-3-one, 3-oxo-1,2-indazole, 3h-indazol-3-one, 1,2-dihydro, 2h-indazol-3-ol
C1=CC=C2C(=C1)C(=O)NN2
134.138
134.14
≥98.0% (T)
250°C
7364-25-2
MFCD00005685
SWEICGMKXPNXNU-UHFFFAOYSA-N
1,2-dihydroindazol-3-one
81829
Crystal-Powder at 20°C
3-Indazolinone
Safety and Handling

Safety and Handling

EINECSNumber : (5)-0269

TSCA : Yes