missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
3-Indazolinone, 97%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals L0586006
Description
3-Indazolinone is used as catalytic agent, petrochemical additive.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications3-Indazolinone is used as catalytic agent, petrochemical additive.
Notes
Store in a cool, dry conditions in well sealed container. Incompatible with oxidizing agents.
Specifications
3-Indazolinone | |
248°C to 252°C | |
MFCD00005685 | |
3733 | |
SWEICGMKXPNXNU-UHFFFAOYSA-N | |
1,2-dihydroindazol-3-one | |
81829 | |
97% |
7364-25-2 | |
C7H6N2O | |
5 g | |
1h-indazol-3-ol, 3-indazolinone, indazolinone, 1h-indazol-3 2h-one, 3-hydroxy-1h-indazole, 3-hydroxyindazole, 1,2-dihydro-3h-indazol-3-one, 3-oxo-1,2-indazole, 3h-indazol-3-one, 1,2-dihydro, 2h-indazol-3-ol | |
C1=CC=C2C(=C1)C(=O)NN2 | |
134.138 | |
134.14 |
Chemical Identifiers
7364-25-2 | |
134.138 | |
SWEICGMKXPNXNU-UHFFFAOYSA-N | |
81829 | |
C1=CC=C2C(=C1)C(=O)NN2 |
C7H6N2O | |
MFCD00005685 | |
1h-indazol-3-ol, 3-indazolinone, indazolinone, 1h-indazol-3 2h-one, 3-hydroxy-1h-indazole, 3-hydroxyindazole, 1,2-dihydro-3h-indazol-3-one, 3-oxo-1,2-indazole, 3h-indazol-3-one, 1,2-dihydro, 2h-indazol-3-ol | |
1,2-dihydroindazol-3-one |
Safety and Handling
EINECSNumber : 230-904-2
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only