Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Hydroxyxanthone, 98%

Chemical Identifiers

Chemical Name or Material 3-Hydroxyxanthone
CAS 3722-51-8
Formula Weight 212.2
MDL Number MFCD00604748
Beilstein 170775
Synonym 3-Hydroxyxanthen-9-one; Sieber linker
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6349503

  SDS
alfa aesar™
H6349503
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6349506

  SDS
alfa aesar™
H6349506
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 212.2
Chemical Name or Material 3-Hydroxyxanthone
Physical Form Solid
Assay 98%
MDL Number MFCD00604748
CAS 3722-51-8
Beilstein 170775
Synonym 3-Hydroxyxanthen-9-one; Sieber linker
Storage Note 1 -30° to -10°C
Melting Point 243°C (dec.)
Quantity 1g
Safety and Handling

-30° to -10°C