missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Hydroxyxanthone, 98%

Manufacturer:  Alfa Aesar™ H6349506

 View more versions of this product

Catalog No. AAH6349506



Specifications

Specifications

3-Hydroxyxanthone
5g
C13H8O3
170775
XCJHDJAODLKGLG-UHFFFAOYSA-N
3-hydroxyxanthen-9-one
5376013
Solid
243°C (decomposition)
3722-51-8
MFCD00604748
sieber linker, 3-hydroxy-9h-xanthen-9-one, 3-hydroxy-xanthen-9-one, 3-hydroxyxanthone, 9h-xanthen-9-one, 3-hydroxy, sieber linker;, xanthen-9-one, 3-hydroxy, pubchem12819, acmc-209irq, 3-hydroxy-9-xanthenone
C1=CC=C2C(=C1)C(=O)C3=C(O2)C=C(C=C3)O
212.204
212.2
98%

Chemical Identifiers

3722-51-8
212.204
XCJHDJAODLKGLG-UHFFFAOYSA-N
5376013
C1=CC=C2C(=C1)C(=O)C3=C(O2)C=C(C=C3)O
C13H8O3
MFCD00604748
sieber linker, 3-hydroxy-9h-xanthen-9-one, 3-hydroxy-xanthen-9-one, 3-hydroxyxanthone, 9h-xanthen-9-one, 3-hydroxy, sieber linker;, xanthen-9-one, 3-hydroxy, pubchem12819, acmc-209irq, 3-hydroxy-9-xanthenone
3-hydroxyxanthen-9-one
Safety and Handling

Safety and Handling

storageNote1 : -30° to -10°C

SDS Documents